Alfa Aesar™ Tris(cyclopentadienyl)gadolinium(III), 99.9% (REO)

Manufacturer: Alfa Aesar™ 3858503

Quantity 1g

 View more versions of this product

Catalog No. AA3858503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1272-21-5
    Formula Weight 352.54
    MDL Number MFCD00145476
    UN Number UN3181
    Chemical Name or Material Tris(cyclopentadienyl)gadolinium(III)

    Specifications

    Formula Weight 352.54
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.
    CAS 1272-21-5
    UN Number UN3181
    Health Hazard 1 Warning
    Sensitivity Air & Moisture Sensitive
    Physical Form Powder
    MDL Number MFCD00145476
    Storage Note 1
    • Ampouled under argon
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    Quantity 1g
    Chemical Name or Material Tris(cyclopentadienyl)gadolinium(III)
    Assay 99.9%, (REO)

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit