Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tris DBA, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
N-myristoyltransferase-1 (NMT-1) inhibitor that prevents activation of MAPK, PI 3-K and STAT3 signaling pathways. Exhibits antiproliferative activity against melanoma cells in vitro and in vivo. Also used as a cyclization catalyst.
Specifications
Quantity | 10 mg |
Formula Weight | 915.73 |
Percent Purity | >98% |
Chemical Name or Material | Tris DBA |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction