Alfa Aesar™ Tris(dibenzoylmethane)mono(phenanthroline)europium(III)

As an organic red light emitter

Manufacturer: Alfa Aesar™ 4417502

Quantity 0.5g

 View more versions of this product

Catalog No. AA4417502

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17904-83-5
    Molecular Formula C57H41EuN2O6
    Formula Weight 1001.93
    MDL Number MFCD01321202
    Chemical Name or Material Tris (dibenzoylmethane)mono (phenanthroline)europium (III)

    Specifications

    Formula Weight 1001.93
    CAS 17904-83-5
    Physical Form Powder
    Molecular Formula C57H41EuN2O6
    MDL Number MFCD01321202
    Solubility Information Soluble in dichloromethane, THF
    Melting Point 172° to 173°C
    Quantity 0.5g
    Chemical Name or Material Tris (dibenzoylmethane)mono (phenanthroline)europium (III)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit