Tris(dibenzylideneacetone)dipalladium(0), Pd 21.5% min., Alfa Aesar™

Manufacturer: Alfa Aesar 1276003

Quantity 1g

 View more versions of this product

Catalog No. AA1276003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 51364-51-3
    Physical Form Powder
    Molecular Formula C51H42O3Pd2
    Formula Weight 915.74
    MDL Number MFCD00013310
    Synonym Bis[tris(dibenzylideneacetone)palladium(0)]
    Chemical Name or Material Tris(dibenzylideneacetone)dipalladium(0)

    Specifications

    Formula Weight 915.74
    Quantity 1g
    Physical Form Powder
    Molecular Formula C51H42O3Pd2
    MDL Number MFCD00013310
    CAS 51364-51-3
    Synonym Bis[tris(dibenzylideneacetone)palladium(0)]
    Chemical Name or Material Tris(dibenzylideneacetone)dipalladium(0)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit