Alfa Aesar™ Tris(diethylamino)gallium(III), 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 42759ME

Quantity 500mg

 View more versions of this product

Catalog No. AA42759ME

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 194611-64-8
    Molecular Formula C12H30GaN3
    Linear Formula Ga[N(CH2CH3)2]3
    Formula Weight 286.11
    MDL Number MFCD01863475
    UN Number UN3263
    Synonym Gallium diethylamide
    Chemical Name or Material Tris (diethylamino)gallium (III)

    Specifications

    Formula Weight 286.11
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P402
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Synonym Gallium diethylamide
    Molecular Formula C12H30GaN3
    MDL Number MFCD01863475
    CAS 194611-64-8
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
    Sensitivity Moisture Sensitive
    Physical Form Powder
    Linear Formula Ga[N(CH2CH3)2]3
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Color Pale brown
    Quantity 500mg
    UN Number UN3263
    Health Hazard 1 Danger
    Chemical Name or Material Tris (diethylamino)gallium (III)
    Assay 99.9%, (metals basis)

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P402
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit