Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tris(diethylamino)gallium(III), 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4275904

Quantity 2g

 View more versions of this product

Catalog No. AA4275904


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

194611-64-8
Ga[N(CH2CH3)2]3
MFCD01863475
Gallium diethylamide
C12H30GaN3
286.11
UN3263
Tris (diethylamino)gallium (III)

Specifications

Moisture Sensitive
194611-64-8
C12H30GaN3
286.11
UN3263
Pale brown
99.9%, (metals basis)
2g
Powder
Ga[N(CH2CH3)2]3
MFCD01863475
Gallium diethylamide
Tris (diethylamino)gallium (III)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit