Alfa Aesar™ Tris(dimethylamino)aluminum dimer

Preparation of aluminum nitrides

Manufacturer: Alfa Aesar™ 4323706

Quantity 5g

 View more versions of this product

Catalog No. AA4323706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32093-39-3
    Molecular Formula C12H36Al2N6
    Linear Formula Al2[N(CH3)2]6
    Formula Weight 318.42
    MDL Number MFCD00269811
    UN Number UN3131
    Synonym Aluminum dimethylamide; Hexakisdimethylamide dialuminum
    Chemical Name or Material Tris (dimethylamino)aluminum dimer

    Specifications

    Formula Weight 318.42
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.
    Physical Form Powder
    Linear Formula Al2[N(CH3)2]6
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228-H260-H314-H318
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Boiling Point 90°C (0.05mmHg) (subl.)
    CAS 32093-39-3
    Flash Point 21°C (70°F)
    Storage Note 1
    • Packaged under Argon
    Sensitivity Moisture Sensitive
    MDL Number MFCD00269811
    UN Number UN3131
    Synonym Aluminum dimethylamide; Hexakisdimethylamide dialuminum
    Density 0.865g/mL
    Color White to pale brown
    Melting Point 87° to 89°C
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
    Molecular Formula C12H36Al2N6
    Quantity 5g
    Chemical Name or Material Tris (dimethylamino)aluminum dimer

    GHS H Statement
    H228-H260-H314-H318
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit