Alfa Aesar™ Tris(dimethylamino)gallium(III) dimer, 99.9% (metals basis)

Gallium nitride film precursor

Manufacturer: Alfa Aesar™ 4044327

Quantity 2.5g

 View more versions of this product

Catalog No. AA4044327

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 57731-40-5
    Linear Formula Ga2[N(CH3)2]6
    Formula Weight 403.9
    MDL Number MFCD00798535
    UN Number UN3131
    Synonym Gallium dimethylamide
    Chemical Name or Material Tris(dimethylamino)gallium(III) dimer

    Specifications

    Physical Form Powder/Chunks
    Formula Weight 403.9
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.
    Health Hazard 2 GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    CAS 57731-40-5
    UN Number UN3131
    Quantity 2.5g
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
    Melting Point 91°C
    Synonym Gallium dimethylamide
    Storage Note 1
    • Ampouled under argon
    Color White to yellow
    Boiling Point 125°C (0.01mmHg) (subl.)
    Sensitivity Moisture Sensitive
    Linear Formula Ga2[N(CH3)2]6
    MDL Number MFCD00798535
    Health Hazard 1 Danger
    Chemical Name or Material Tris(dimethylamino)gallium(III) dimer
    Assay 99.9%, (metals basis)

    GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit