Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(dimethylamino)methane, 85%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
5762-56-1
C7H19N3
145.25
MFCD00008322
MUMVIYLVHVCYGI-UHFFFAOYSA-N
tris dimethylamino methane, n,n,n',n',n,n-hexamethylmethanetriamine, methanetriamine, n,n,n',n',n,n-hexamethyl, bis dimethylamino methyl dimethylamine, n,n,n,n,n,n-hexamethylmethanetriamine, trisdimethylaminomethane, acmc-209lzg, tris-dimethylaminomethane
79831
N,N,N',N',N'',N''-hexamethylmethanetriamine
CN(C)C(N(C)C)N(C)C
5762-56-1
C7H19N3
145.25
MFCD00008322
MUMVIYLVHVCYGI-UHFFFAOYSA-N
tris dimethylamino methane, n,n,n',n',n,n-hexamethylmethanetriamine, methanetriamine, n,n,n',n',n,n-hexamethyl, bis dimethylamino methyl dimethylamine, n,n,n,n,n,n-hexamethylmethanetriamine, trisdimethylaminomethane, acmc-209lzg, tris-dimethylaminomethane
79831
N,N,N',N',N'',N''-hexamethylmethanetriamine
CN(C)C(N(C)C)N(C)C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Density 0.8380g/mL
Quantity 5g
Assay Percent Range 85%
Linear Formula CH[N(CH3)2]3
Formula Weight 145.25
Physical Form Liquid
Percent Purity ≥80%
Chemical Name or Material Tris(dimethylamino)methane
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Extremely flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger