Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TE Buffer, Tris-EDTA, 100X Powder, pH 8.0, Molecular Biology, Fisher BioReagents

Catalog No. BP1339-1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Save time and simplify your preparation of buffer concentrate by using Fisher BioReagents 100X TE powder - simply add pure water
  • Each pack contains the correct amount of pre-weighed powder to make 1L of 100X TE buffer containing 1.0 M tris and 0.1M EDTA
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer is routinely used for resuspending nucleic acid samples
  • Intended for laboratory research use only
Specifications

Specifications

Soluble in water
100X Powder
560 to 740μmhos/cm
DNase free
Molecular Biology
Protease free
White
Tris-EDTA
77-86-1, 60-00-4
TE
Solid
Foil Pack
170.2 to 175.4g
1 pk.
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Causes eye, skin, and respiratory tract irritation. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:0

Recommended Storage : RT

SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit