Learn More
TE Buffer, Tris-EDTA, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents
Description
- Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TE buffer solution - simply dilute as needed
- 10X solution contains 100mM tris and 10mM EDTA
- To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TE solution with 9 volumes of ultrapure water, such as BP2819
- Filtered through a 5 micron filter and autoclaved
- Tested for the absence of DNase, RNase, and protease
- TE buffer is routinely used for resuspending nucleic acid samples
- Intended for laboratory research use only
Specifications
Tris-EDTA (TE), 10X Solution | |
77-86-1, 6381-92-6, 7647-01-0, 7732-18-5 | |
7.4 to 8.1 | |
500 mL | |
Filtered through a 5-micron filter and autoclaved. | |
Protease free | |
Molecular Biology |
pH 7.4 | |
Colorless | |
Liquid | |
DNase free | |
Poly Bottle | |
TE |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.
NFPA
Health:1
Flammability:0
Instability:0
Recommended Storage : RT
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.