Alfa Aesar™ Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate

Manufacturer: Alfa Aesar™ 2412706

Quantity 5g

 View more versions of this product

Catalog No. AA2412706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16165-32-5
    Linear Formula Cr(H2NCH2CH2NH2)3Cl3·3.5H2O
    Formula Weight 401.71 (338.65 Anhydrous)
    MDL Number MFCD00150160
    Synonym Trichlorotris(ethylenediamine)chromium(III)
    Chemical Name or Material Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate

    Specifications

    Formula Weight 401.71 (338.65 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    Synonym Trichlorotris(ethylenediamine)chromium(III)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    Linear Formula Cr(H2NCH2CH2NH2)3Cl3·3.5H2O
    MDL Number MFCD00150160
    CAS 16165-32-5
    Color Yellow to orange
    Quantity 5g
    Chemical Name or Material Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit