Alfa Aesar™ Tris(ethylenediamine)iron(III) sulfate

Manufacturer: Alfa Aesar™ 3112318

Quantity 50g

 View more versions of this product

Catalog No. AA3112318

  Check Availability
Add to Cart

    Chemical Identifiers

    Formula Weight 760.38
    MDL Number MFCD00035682
    Chemical Name or Material Tris(ethylenediamine)iron(III) sulfate

    Specifications

    Formula Weight 760.38
    MDL Number MFCD00035682
    Physical Form Powder
    Quantity 50g
    Chemical Name or Material Tris(ethylenediamine)iron(III) sulfate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit