Alfa Aesar™ Tris(ethylenediamine)rhodium(III) chloride trihydrate, Rh 23.2%

Manufacturer: Alfa Aesar™ 1055302

Quantity 0.5g

 View more versions of this product

Catalog No. AA1055302

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15004-86-1
    Linear Formula Rh(H2NCH2CH2NH2)3Cl3·3H2O
    Formula Weight 443.61 (389.56 Anhydrous)
    MDL Number MFCD00149661
    Synonym Trichlorotris(ethylenediamine)rhodium(III)
    Chemical Name or Material Tris(ethylenediamine)rhodium(III) chloride trihydrate

    Specifications

    Physical Form Crystalline/Fused Lump, Crystals Fuse Over Time
    Formula Weight 443.61 (389.56 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00149661
    Synonym Trichlorotris(ethylenediamine)rhodium(III)
    Health Hazard 1 Warning
    Color White
    Melting Point 280°C (dec.)
    CAS 15004-86-1
    Concentration Rh 23.2%
    Quantity 0.5g
    Linear Formula Rh(H2NCH2CH2NH2)3Cl3·3H2O
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Tris(ethylenediamine)rhodium(III) chloride trihydrate

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit