Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tris Hydrochloride, Molecular Biology Grade, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
500g
1kg
5kg
25kg
Packaging:
Plastic bottle
1185-53-1
C4H12ClNO3
157.594
MFCD00012590
QKNYBSVHEMOAJP-UHFFFAOYSA-N
Tris(hydroxymethyl)amino methane hydrochloride
93573
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
C(C(CO)(CO)N)O.Cl
1185-53-1
C4H12ClNO3
157.594
MFCD00012590
QKNYBSVHEMOAJP-UHFFFAOYSA-N
Tris(hydroxymethyl)amino methane hydrochloride
93573
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
C(C(CO)(CO)N)O.Cl
Formula Weight 157.60
Physical Form Crystalline Powder
Grade Molecular Biology
Packaging Plastic bottle
Melting Point ~152°C (decomposition)
Quantity 5kg
Chemical Name or Material Tris hydrochloride