Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS Hydrochloride, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
250g
500g
1kg
5kg
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

1185-53-1
C4H12ClNO3
157.594
QKNYBSVHEMOAJP-UHFFFAOYSA-N
93573
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
C(C(CO)(CO)N)O.Cl

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology Applications
Packaging Poly Bottle
Formula Weight 157.6
Quantity 1kg
Chemical Name or Material TRIS Hydrochloride
Grade Ultrapure Bioreagent