Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(hydroxymethyl)aminomethane, ACS Reagent (Tris; THAM; 2-Amino-2-(hydroxymethyl)-1,3-propanediol), Reagents

Catalog No. 01337635
Click to view available options
Quantity:
100 g
500 g
5 kg

Tris(hydroxymethyl)aminomethane, ACS Reagent (Tris; THAM; 2-Amino-2-(hydroxymethyl)-1,3-propanediol), ACS, use for Buffers, Acid Titrations. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Tromethamine
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES NC(CO)(CO)CO

Specifications

Melting Point 171°C to 172°C
Boiling Point 219°C
Color White
For Use With (Application) Titration
For Use With (Equipment) pH Meter
Physical Form Solid
Quantity 5 kg
Packaging Poly Pail
Specific Gravity 1.35
Sterility Non-sterile
Grade ACS
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.