Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(hydroxymethyl)phosphine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
25g
Packaging:
Glass bottle
2767-80-8
C3H9O3P
124.08
MFCD00055382
JMXMXKRNIYCNRV-UHFFFAOYSA-N
tris hydroxymethyl phosphine,methanol, phosphinidynetris,trimethylolphosphine,phosphinidynetrimethanol,tris methanol phosphine,phosphinidynetrismethanol,unii-y6tg7wf7oq,y6tg7wf7oq,methanol, 1,1',1-phosphinidynetris,methanol, phosphinidynetri-7ci,8ci
76001
bis(hydroxymethyl)phosphanylmethanol
C(O)P(CO)CO
2767-80-8
C3H9O3P
124.08
MFCD00055382
JMXMXKRNIYCNRV-UHFFFAOYSA-N
tris hydroxymethyl phosphine,methanol, phosphinidynetris,trimethylolphosphine,phosphinidynetrimethanol,tris methanol phosphine,phosphinidynetrismethanol,unii-y6tg7wf7oq,y6tg7wf7oq,methanol, 1,1',1-phosphinidynetris,methanol, phosphinidynetri-7ci,8ci
76001
bis(hydroxymethyl)phosphanylmethanol
C(O)P(CO)CO
Melting Point 48°C to 56°C
Boiling Point 111.0°C to 113.0°C (2.5 mmHg)
Color White to Yellow
NMR 94% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Flash Point >110°C
Assay Percent Range 95%
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 124.08
Physical Form Powder, Crystals or Chunks
Percent Purity 95%
Chemical Name or Material Tris(hydroxymethyl)phosphine
Show More Show Less

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 220-445-6