Alfa Aesar™ Tris(isopropylcyclopentadienyl)terbium(III), 99.9% (REO)

Manufacturer: Alfa Aesar™ 4050603

Quantity 1g

 View more versions of this product

Catalog No. AA4050603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 312696-25-6
    Formula Weight 480.45
    MDL Number MFCD00798537
    UN Number UN3181
    Chemical Name or Material Tris(isopropylcyclopentadienyl)terbium(III)

    Specifications

    Formula Weight 480.45
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 1g
    Physical Form Crystalline
    MDL Number MFCD00798537
    Health Hazard 1 Warning
    Sensitivity Air & Moisture Sensitive
    Storage Note 1
    • Ampouled under argon
    CAS 312696-25-6
    UN Number UN3181
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Chemical Name or Material Tris(isopropylcyclopentadienyl)terbium(III)
    Assay 99.9%, (REO)

    GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit