Alfa Aesar™ Tris(n-butylcyclopentadienyl)yttrium(III), 99.9% (REO)

Manufacturer: Alfa Aesar™ 3957703

Quantity 1g

 View more versions of this product

Catalog No. AA3957703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 312739-77-8
    Formula Weight 452.52
    MDL Number MFCD00145473
    UN Number UN1993
    Chemical Name or Material Tris(n-butylcyclopentadienyl)yttrium(III)

    Specifications

    Physical Form Liquid, Ampouled Under Argon
    Formula Weight 452.52
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00145473
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Quantity 1g
    CAS 312739-77-8
    UN Number UN1993
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material Tris(n-butylcyclopentadienyl)yttrium(III)
    Assay 99.9%, (REO)

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit