Alfa Aesar™ Tris[N,N-bis(trimethylsilyl)amide]cerium(III), 98%

Manufacturer: Alfa Aesar™ 4436206

Quantity 5g

 View more versions of this product

Catalog No. AA4436206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 41836-21-9
    Molecular Formula C18H54CeN3Si6
    Linear Formula [[(CH3)3Si]2N]3Ce
    Formula Weight 621.27
    MDL Number MFCD03411241
    UN Number UN3396
    Synonym Cerium(III) tris[N,N-bis(trimethylsilyl)amide]
    Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]cerium (III)

    Specifications

    Formula Weight 621.27
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.
    Linear Formula [[(CH3)3Si]2N]3Ce
    MDL Number MFCD03411241
    Synonym Cerium(III) tris[N,N-bis(trimethylsilyl)amide]
    Physical Form Powder
    Molecular Formula C18H54CeN3Si6
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE
    CAS 41836-21-9
    UN Number UN3396
    Health Hazard 2 GHS H Statement
    H261-H228-H314-H318
    In contact with water releases flammable gas.
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Color Bright yellow
    Quantity 5g
    Sensitivity Moisture Sensitive
    Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]cerium (III)
    Assay 98%

    GHS H Statement
    H261-H228-H314-H318
    In contact with water releases flammable gas.
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit