Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris[N,N-bis(trimethylsilyl)amide]erbium(III), 98%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
103457-72-3
C18H54ErN3Si6
648.42
MFCD00271002
ZVNGWTZZRANJAO-UHFFFAOYSA-N
bis trimethylsilyl azanide; erbium 3+,tris n,n-bis trimethylsilyl amide erbium iii,acmc-20al2x,erbium iii bis trimethylsilyl amide,tris bis trimethylsilyl amino erbium,erbium tris bis trimethylsilyl amide,erbium 3+ tris bis trimethylsilyl azanide,erbium tris trimethyl-n-trimethylsilyl silanaminide,silanamine,1,1,1-trimethyl-n-trimethylsilyl-, erbium 3+ salt 3:1
6098739
bis(trimethylsilyl)azanide;erbium(3+)
C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Er+3]
103457-72-3
C18H54ErN3Si6
648.42
MFCD00271002
ZVNGWTZZRANJAO-UHFFFAOYSA-N
bis trimethylsilyl azanide; erbium 3+,tris n,n-bis trimethylsilyl amide erbium iii,acmc-20al2x,erbium iii bis trimethylsilyl amide,tris bis trimethylsilyl amino erbium,erbium tris bis trimethylsilyl amide,erbium 3+ tris bis trimethylsilyl azanide,erbium tris trimethyl-n-trimethylsilyl silanaminide,silanamine,1,1,1-trimethyl-n-trimethylsilyl-, erbium 3+ salt 3:1
6098739
bis(trimethylsilyl)azanide;erbium(3+)
C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Er+3]
Melting Point 180°C to 184°C
Color Yellow to Orange
Quantity 1g
Elemental Analysis 32.5 to 34 % (C) 8.2 to 8.6 % (H)
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula Er[N(Si(CH3)3)2]3
Solubility Information Solubility in water: insoluble
Formula Weight 648.42
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Tris[N, N-bis(trimethylsilyl)amide]erbium(III)
Show More Show Less

GHS H Statement
Flammable solid.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.
Reacts violently with water.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye prot

GHS Signal Word: Danger