Alfa Aesar™ Tris[N,N-bis(trimethylsilyl)amide]europium(III), 98%

Manufacturer: Alfa Aesar™ 4437603

Quantity 1g

 View more versions of this product

Catalog No. AA4437603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 35789-02-7
    Molecular Formula C18H54EuN3Si6
    Linear Formula [[(CH3)3Si]2N]3Eu
    Formula Weight 633.13
    MDL Number MFCD03411248
    UN Number UN3396
    Synonym Europium(III) tris[N,N-bis(trimethylsilyl)amide]
    Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]europium (III)

    Specifications

    Formula Weight 633.13
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.
    CAS 35789-02-7
    Melting Point 180° to 184°C
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE
    Synonym Europium(III) tris[N,N-bis(trimethylsilyl)amide]
    Boiling Point 105°C (104mmHg)
    Sensitivity Moisture Sensitive
    Molecular Formula C18H54EuN3Si6
    Linear Formula [[(CH3)3Si]2N]3Eu
    UN Number UN3396
    Quantity 1g
    Physical Form Powder
    MDL Number MFCD03411248
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H261-H228-H314-H318
    In contact with water releases flammable gas.
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Color Orange
    Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]europium (III)
    Assay 98%

    GHS H Statement
    H261-H228-H314-H318
    In contact with water releases flammable gas.
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit