Alfa Aesar™ Tris[N,N-bis(trimethylsilyl)amide]gadolinium(III), 98%

Manufacturer: Alfa Aesar™ 4435903

Quantity 1g

 View more versions of this product

Catalog No. AA4435903

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 35789-03-8
    Molecular Formula C18H54GdN3Si6
    Linear Formula [[(CH3)3Si]2N]3Gd
    Formula Weight 638.41
    MDL Number MFCD03427091
    UN Number UN3396
    Synonym Gadolinium(III) tris[N,N-bis(trimethylsilyl)amide]
    Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]gadolinium (III)

    Specifications

    Formula Weight 638.41
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.
    UN Number UN3396
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE
    Molecular Formula C18H54GdN3Si6
    Linear Formula [[(CH3)3Si]2N]3Gd
    Synonym Gadolinium(III) tris[N,N-bis(trimethylsilyl)amide]
    Melting Point 161° to 167°C
    Quantity 1g
    Sensitivity Moisture Sensitive
    Physical Form Powder
    CAS 35789-03-8
    MDL Number MFCD03427091
    Health Hazard 2 GHS H Statement
    H261-H228-H314-H318
    In contact with water releases flammable gas.
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]gadolinium (III)
    Assay 98%

    GHS H Statement
    H261-H228-H314-H318
    In contact with water releases flammable gas.
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit