Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tris[N,N-bis(trimethylsilyl)amide]neodymium(III), 98%

Manufacturer:  Alfa Aesar™ 4439503

Quantity 1g

 View more versions of this product

Catalog No. AA4439503

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

148274-47-9
[[(CH3)3Si]2N]3Nd
MFCD03427089
Neodymium(III) tris[N,N-bis(trimethylsilyl)amide]
C18H54N3NdSi6
625.39
UN3396
Tris[N,N-bis (trimethylsilyl)amide]neodymium (III)

Specifications

148274-47-9
C18H54N3NdSi6
625.39
UN3396
Pale purple
1g
Tris[N,N-bis (trimethylsilyl)amide]neodymium (III)
Powder
[[(CH3)3Si]2N]3Nd
MFCD03427089
Neodymium(III) tris[N,N-bis(trimethylsilyl)amide]
156° to 159°C
Moisture Sensitive
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H261-H228-H314-H318
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit