Alfa Aesar™ Tris[N,N-bis(trimethylsilyl)amide]yttrium(III), 98+%

Manufacturer: Alfa Aesar™ 3957803

Quantity 1g

 View more versions of this product

Catalog No. AA3957803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 41836-28-6
    Linear Formula [[(CH3)3Si]2N]3Y
    Formula Weight 570.06
    MDL Number MFCD00210649
    UN Number UN3396
    Synonym Yttrium(III) tris[N,N-bis(trimethylsilyl)amide]
    Chemical Name or Material Tris[N,N-bis(trimethylsilyl)amide]yttrium(III)

    Specifications

    Formula Weight 570.06
    Boiling Point 105°C (at 10-4mmHg) (subl.)
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.
    CAS 41836-28-6
    Health Hazard 2 GHS H Statement
    H261-H228-H314-H318
    In contact with water releases flammable gas.
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    MDL Number MFCD00210649
    Synonym Yttrium(III) tris[N,N-bis(trimethylsilyl)amide]
    Health Hazard 1 Danger
    Color White
    Quantity 1g
    Sensitivity Moisture Sensitive
    UN Number UN3396
    Melting Point 180° to 184°C
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE
    Physical Form Powder
    Linear Formula [[(CH3)3Si]2N]3Y
    Storage Note 1
    • Ampouled under argon
    Chemical Name or Material Tris[N,N-bis(trimethylsilyl)amide]yttrium(III)
    Assay ≥98%

    GHS H Statement
    H261-H228-H314-H318
    In contact with water releases flammable gas.
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit