Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris Solution, 1.0M, pH 8.0, OmniPur™, Calbiochem™,

Click to view available options
Quantity:
100mL
Packaging:
Plastic bottle
77-86-1,77-86-1,7647-01-0
1M Solution
C4H11NO3
Clear, colorless liquid
Tris Solution
77-86-1,77-86-1,7647-01-0
1M Solution
C4H11NO3
Clear, colorless liquid
Tris Solution

Useful in the pH range of 8.0

Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Sterility Passes Test
DNase None Detected
Solubility Information Soluble in water
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Molecular Weight (g/mol) 121.14
PubChem CID 6503
ChEBI CHEBI:9754
Grade Molecular Biology
Packaging Plastic bottle
pH Range pH of solution at 25°C: 7.9-8.1
Protease None Detected
RNase None Detected
Concentration or Composition Notes Molarity: 0.9-1.1 M
RNase, DNase, Nickase, and Protease None detected
Quantity 100mL
Show More Show Less

Recommended Storage : Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.