Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(tetramethylcyclopentadienyl)cerium, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Ampule in Plastic Container
251984-08-4
C27H39Ce
503.725
MFCD01863662
VNYOSWSTNAVLMJ-UHFFFAOYSA-N
tris tetramethylcyclopentadienyl cerium iii,tris 1,2,3,4-tetramethyl-2,4-cyclopentadienyl cerium iii
121233288
cerium(3+);1,2,3,5-tetramethylcyclopenta-1,3-diene
C[C-]1C=C(C(=C1C)C)C.C[C-]1C=C(C(=C1C)C)C.C[C-]1C=C(C(=C1C)C)C.[Ce+3]
251984-08-4
C27H39Ce
503.725
MFCD01863662
VNYOSWSTNAVLMJ-UHFFFAOYSA-N
tris tetramethylcyclopentadienyl cerium iii,tris 1,2,3,4-tetramethyl-2,4-cyclopentadienyl cerium iii
121233288
cerium(3+);1,2,3,5-tetramethylcyclopenta-1,3-diene
C[C-]1C=C(C(=C1C)C)C.C[C-]1C=C(C(=C1C)C)C.C[C-]1C=C(C(=C1C)C)C.[Ce+3]
Packaging Ampule in Plastic Container
Quantity 1g
NMR Complies
Color Green
Formula Weight 503.73
Physical Form Needles
Chemical Name or Material Tris(tetramethylcyclopentadienyl)cerium

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning