Learn More
TRIS-Tricine-SDS running buffer (10X), anode buffer, pH 8.9, Thermo Scientific™
Specifications
Chemical Name or Material | TRIS-Tricine-SDS running buffer (10X) |
---|---|
Physical Form | Liquid |
Packaging | Plastic bottle |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ63375K2
|
Thermo Scientific Chemicals
J63375K2 |
1 L | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
AAJ63375AP
|
Thermo Scientific Chemicals
J63375AP |
500 mL | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
TRIS-Tricine-SDS running buffer (10X) | |
Liquid | |
Plastic bottle |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.