Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(trifluoro-2,4-pentanedionato)aluminum(III) 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
14354-59-7
C15H15AlF9O6
489.25
MFCD00042062
DOYKOKXTRGISGV-DJFUMVPSSA-N
Aluminum(III) Trifluoroacetylacetonate, Trifluoroacetylacetonato Aluminum(III) Salt
14354-59-7
C15H15AlF9O6
489.25
MFCD00042062
DOYKOKXTRGISGV-DJFUMVPSSA-N
Aluminum(III) Trifluoroacetylacetonate, Trifluoroacetylacetonato Aluminum(III) Salt
Color Undesignated
Quantity 25g
Melting Point 121°C
Formula Weight 486.22
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Tris(trifluoro-2,4-pentanedionato)aluminum(III)