Learn More
Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III), Thermo Scientific Chemicals
Description
Optically active NMR shift reagent. Used with Ag(fod) for the enantiomeric resolution of sec-butylisothiouronium chloride. Also reactant involved in synthesis of europium acetyl camphorate phosphine oxide complexes and reactions with 1,10-N,N-phenanthroline dioxide derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 195°C to 198°C |
Quantity | 5 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in chloroform. Insoluble in water. |
Formula Weight | 893.68 |
Physical Form | Crystalline |
Chemical Name or Material | Tris (trifluoromethylhydroxymethylene-d-camphorato)europium (III) |
Safety and Handling
EINECSNumber | 252-232-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.