Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(trimethylsilyl)amine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1586-73-8
C9H27NSi3
233.58
MFCD00047990
PEGHITPVRNZWSI-UHFFFAOYSA-N
tris trimethylsilyl amine,amine, tris trimethylsilyl,silanamine, 1,1,1-trimethyl-n,n-bis trimethylsilyl,silanamine,1,1,1-trimethyl-n,n-bis trimethylsilyl,nitrilotris trimethylsilane,ch3 3si 3n,n,n,n-tris trimethylsilyl amine #,bis trimethylsilyl amino-dimethylsilyl methane
74110
[[bis(trimethylsilyl)amino]-dimethylsilyl]methane
C[Si](C)(C)N([Si](C)(C)C)[Si](C)(C)C
1586-73-8
C9H27NSi3
233.58
MFCD00047990
PEGHITPVRNZWSI-UHFFFAOYSA-N
tris trimethylsilyl amine,amine, tris trimethylsilyl,silanamine, 1,1,1-trimethyl-n,n-bis trimethylsilyl,silanamine,1,1,1-trimethyl-n,n-bis trimethylsilyl,nitrilotris trimethylsilane,ch3 3si 3n,n,n,n-tris trimethylsilyl amine #,bis trimethylsilyl amino-dimethylsilyl methane
74110
[[bis(trimethylsilyl)amino]-dimethylsilyl]methane
C[Si](C)(C)N([Si](C)(C)C)[Si](C)(C)C
Boiling Point 72°C (10.0mmHg)
Melting Point 67°C to 69°C
Infrared Spectrum Authentic
Quantity 5g
Color Colorless
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula N(Si(CH3)3)3
Solubility Information Solubility in water: insoluble
Formula Weight 233.58
Physical Form Crystalline
Percent Purity ≥98.5%
Chemical Name or Material Tris(trimethylsilyl)amine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger