Alfa Aesar™ Tris(trimethylsilyl)phosphine

Manufacturer: Alfa Aesar™ 30218MD

Quantity 250mg

 View more versions of this product

Catalog No. AA30218MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15573-38-3
    Formula Weight 250.55
    MDL Number MFCD00015487
    UN Number UN2845
    Chemical Name or Material Tris(trimethylsilyl)phosphine

    Specifications

    Formula Weight 250.55
    Health Hazard 3 GHS P Statement
    P210-P222-P280-P305+P351+P338-P362-P422a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store contents under inert gas.
    MDL Number MFCD00015487
    Quantity 250mg
    UN Number UN2845
    Health Hazard 1 Danger
    Melting Point 24°C
    Boiling Point 102° to 105°C (16mmHg)
    Sensitivity Air & Moisture Sensitive
    Physical Form Liquid
    Refractive Index 1.5020
    Density 0.863g/mL
    Health Hazard 2 GHS H Statement
    H250-H252-H315-H319
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes skin irritation.
    Causes serious eye irritation.
    Odor Pungent
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
    CAS 15573-38-3
    Chemical Name or Material Tris(trimethylsilyl)phosphine

    GHS H Statement
    H250-H252-H315-H319
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P222-P280-P305+P351+P338-P362-P422a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store contents under inert gas.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit