Alfa Aesar™ Tris(trimethylsilyl)silane, 97%

Manufacturer: Alfa Aesar™ B2245714

Quantity 25g

 View more versions of this product

Catalog No. AAB2245714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1873-77-4
    Molecular Formula [(CH3)3Si]3SiH
    Formula Weight 248.67
    MDL Number MFCD00077893
    UN Number UN1993
    Beilstein 1923953
    Merck Index 14,9757
    Synonym TTMSS
    Chemical Name or Material Tris(trimethylsilyl)silane

    Specifications

    Formula Weight 248.67
    UN Number UN1993
    Synonym TTMSS
    Flash Point 55°C (131°F)
    Molecular Formula [(CH3)3Si]3SiH
    MDL Number MFCD00077893
    CAS 1873-77-4
    Beilstein 1923953
    Merck Index 14,9757
    Boiling Point 82° to 84°C (12mmHg)
    Quantity 25g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material Tris(trimethylsilyl)silane
    Assay 97%

    Description

    Applications
    Tris(trimethylsilyl)silane is used in hydrosilylations of carbonyls, radical reactions, reductions of acid chlorides, and carbon-halogen bonds. It acts a mediator of radical reactions and considered to be a nontoxic substitute for tri-n-butylstannane in radical reactions. It forms complexes with transition metals and main group elements. Further, it is utilized in carbon-carbon bond forming reactions.

    Solubility
    Miscible with pentane, ether, toluene and tetrahydrofuran. Sparingly miscible with acetone and acetonitrile. Immiscible with water.

    Notes
    Light sensitive. Incompatible with strong oxidizing agents.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit