Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(triphenylphosphine)rhodium(I) Chloride 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
14694-95-2
C54H45ClP3Rh-
925.231
MFCD00010016
QBERHIJABFXGRZ-UHFFFAOYSA-M
chlorotris triphenylphosphine rhodium i,chlorotris triphenylphosphine rhodium,tris triphenylphosphine rhodium i chloride,tris triphenylphosphine chlororhodium,wilkinson's catalyst,unii-0fv534bkev,tris triphenylphosphine rhodium chloride,0fv534bkev,rhodium; triphenylphosphane; chloride
84599
rhodium;triphenylphosphane;chloride
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Rh]
14694-95-2
C54H45ClP3Rh-
925.231
MFCD00010016
QBERHIJABFXGRZ-UHFFFAOYSA-M
chlorotris triphenylphosphine rhodium i,chlorotris triphenylphosphine rhodium,tris triphenylphosphine rhodium i chloride,tris triphenylphosphine chlororhodium,wilkinson's catalyst,unii-0fv534bkev,tris triphenylphosphine rhodium chloride,0fv534bkev,rhodium; triphenylphosphane; chloride
84599
rhodium;triphenylphosphane;chloride
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Rh]
Color Undesignated
Quantity 5g
Formula Weight 925.23
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Tris(triphenylphosphine)rhodium(I) Chloride