Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(triphenylphosphine)ruthenium(II) chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
15529-49-4
C54H45Cl2P3Ru
958.85
MFCD00013077
WIWBLJMBLGWSIN-UHFFFAOYSA-L
dichlorotris triphenylphosphine ruthenium ii,tris triphenylphosphine ruthenium ii chloride,dichlorotris triphenylphosphino ruthenium ii,tris triphenylphosphine ruthenium ii dichloride,dichlorotris triphenylphosphine ruthenium,ruthenium ii-tris triphenylphosphine dichloride,tris triphenylphosphine dichlororuthenium,ruthenium dichlorotris triphenylphosphine,pubchem12822
11007548
dichlororuthenium;triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl[Ru]Cl
15529-49-4
C54H45Cl2P3Ru
958.85
MFCD00013077
WIWBLJMBLGWSIN-UHFFFAOYSA-L
dichlorotris triphenylphosphine ruthenium ii,tris triphenylphosphine ruthenium ii chloride,dichlorotris triphenylphosphino ruthenium ii,tris triphenylphosphine ruthenium ii dichloride,dichlorotris triphenylphosphine ruthenium,ruthenium ii-tris triphenylphosphine dichloride,tris triphenylphosphine dichlororuthenium,ruthenium dichlorotris triphenylphosphine,pubchem12822
11007548
dichlororuthenium;triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl[Ru]Cl
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Undesignated
Melting Point 159.0°C
Assay Percent Range 98%
Linear Formula [(C6H5)3P]3RuCl2
Fieser 04,564; 05,740; 06,654; 12,179; 16,126; 06,175; 07,101; 08,840; 09,104; 17,33
Solubility Information Solubility in water: insoluble. Other solubilities: sparingly soluble in methanol, ethanol, acetone,, ethyl acetate, chloroform and toluene, insoluble in diethyl ether and n-hexane
Formula Weight 958.85
Physical Form Solution
Percent Purity 98%
Chemical Name or Material Tris(triphenylphosphine)ruthenium(II) chloride
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 239-569-7