Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner TRISMETHOXYRESVERATROL 50 mg  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner B002250MG
Trismethoxyresveratrol (3,5,4'-Trimethoxystilbene) induces microtubule disassembly and tubulin depolimerization. it is also a powerful antiangiogenetic agent and inhibits endothelial cell proliferation, morphogenesis, sprouting and collagen gel invasion.\n\n
\n\n
Documents
\nTechnical Data Sheet
Catalog No. 50-720-003