Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trisodium citrate dihydrate, ACS, 99.0% min, Alfa Aesar™
Click to view available options
Quantity:
500g
2kg
Specifications
Assay Percent Range | 99.0% |
Beilstein | 6104939 |
Solubility Information | Soluble in water. Insoluble in alcohol. |
Formula Weight | 294.10 (258.07 Anhydrous) |
Physical Form | Crystalline |
Grade | ACS Reagent |
Melting Point | 150°C-2H2O |
Quantity | 2kg |
Chemical Name or Material | Trisodium citrate dihydrate, For analysis ACS |
Safety and Handling
H500
EINECSNumber : 200-675-3
TSCA : Yes
Recommended Storage : Ambient temperatures