Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trithiocyanuric acid, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
638-16-4
C3H3N3S3
177.26
MFCD00006052
WZRRRFSJFQTGGB-UHFFFAOYSA-N
trithiocyanuric acid,thiocyanuric acid,1,3,5-triazine-2,4,6-trithiol,trismercaptotriazine,trimercaptocyanuric acid,cyanuric acid, trithio,usaf th-3,1,3,5-triazine-2,4,6 1h,3h,5h-trithione,s-triazine-2,4,6-trithiol,2,4,6-triazinetrithiol
1268121
1,3,5-triazinane-2,4,6-trithione
C1(=S)NC(=S)NC(=S)N1
638-16-4
C3H3N3S3
177.26
MFCD00006052
WZRRRFSJFQTGGB-UHFFFAOYSA-N
trithiocyanuric acid,thiocyanuric acid,1,3,5-triazine-2,4,6-trithiol,trismercaptotriazine,trimercaptocyanuric acid,cyanuric acid, trithio,usaf th-3,1,3,5-triazine-2,4,6 1h,3h,5h-trithione,s-triazine-2,4,6-trithiol,2,4,6-triazinetrithiol
1268121
1,3,5-triazinane-2,4,6-trithione
C1(=S)NC(=S)NC(=S)N1
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Color Yellow
Melting Point >300.0°C
Assay Percent Range 95%
Beilstein 26, 259
Formula Weight 177.26
Physical Form Amorphous Powder
Percent Purity 95%
Chemical Name or Material Trithiocyanuric acid
Show More Show Less

GHS H StatementCauses skin irritation.Causes serious eye irritation.May cause respiratory irritation.

GHS P StatementIF ON SKIN: Wash with plenty of soap and water.If eye irritation persists: Get medical advice/attention.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Call a POISON CENTER or doctor/physician if you feel unwell.Wear protective gloves/protective clothing/eye protection/face protection.If skin irritation occurs: Get medical advice/attention.

;GHS P StatementIF ON SKIN: Wash with plenty of soap and water.If eye irritation persists: Get medical advice/attention.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Call a POISON CENTER or doctor/