Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tritolyl Phosphate, Thermo Scientific™

Click to view available options
Quantity:
1L
Packaging:
EcoSafPak™
1330-78-5
C21H21O4P
MFCD02102116
Tricresyl phosphate
1330-78-5
C21H21O4P
MFCD02102116
Tricresyl phosphate

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Decomposition Information 530°C
Density 12.70
Identification Pass Test
Linear Formula (CH3C6H4O)3PO
Packaging EcoSafPak™
Merck Index 15, 9940
Quantity 1L
Formula Weight 368.36
Vapor Pressure 0.03mmHg at 25°C
Chemical Name or Material Tritolyl phosphate

RUO – Research Use Only