Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triton(R) X-100, cGMP Manufactured, Spectrum™

Click to view available options
Quantity:
1 kg
4 kg
Packaging:
Poly Bottle
9036-19-5
C18H30O3
294.44
LBCZOTMMGHGTPH-UHFFFAOYSA-N
2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethan-1-ol
CC(C)(C)CC(C)(C)C1=CC=C(OCCOCCO)C=C1
9036-19-5
C18H30O3
294.44
LBCZOTMMGHGTPH-UHFFFAOYSA-N
2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethan-1-ol
CC(C)(C)CC(C)(C)C1=CC=C(OCCOCCO)C=C1

T1722, 9036-19-5, C14H21(C2H4O)9-10OH

Spectrum™ Triton(R) X-100, cGMP Manufactured - Spectrum cGMP grade products that have been manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Acid Value 0.2
CAS Max % 1
Dioxane 10 ppm
Ethylene Oxide 5 ppm
Hydroxyl Value 85-101
Ignition Residue 0.004
Packaging Poly Bottle
Quantity 1 kg