Learn More
Research Products International CorpSupplier Diversity Partner Triton X-100, 10% Solution, 5 Ampoules per Pack DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner T600851
Peroxide free detergent in single use ampoules. Ideal for protein isolation and sample preparation. Peroxide content: >1ug/ml. Supplied 5 ampoules per pack. Proteomics Grade
Catalog No. 50-489-120