Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triton™ X-100, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
10L
Packaging:
EcoSafPak™
Glass Bottle
Plastic drum
9002-93-1
C16H26O2
250.382
MFCD00132505
JYCQQPHGFMYQCF-UHFFFAOYSA-N
Polyoxyethylene(10) octylphenyl ether
5590
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
9002-93-1
C16H26O2
250.382
MFCD00132505
JYCQQPHGFMYQCF-UHFFFAOYSA-N
Polyoxyethylene(10) octylphenyl ether
5590
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
Density 1.0670g/mL
Merck Index 15, 6850
Solubility Information Solubility in water: soluble. Other solubilities: soluble in toluene, xylene, trichloroethylene,, ethylene glycol, ethyl ether, ethyl alcohol,, isopropyl alcohol, ethylene dichloride and most, other solvnets., insoluble in kerosene, miniral spirits.
Packaging Plastic drum
Refractive Index 1.4900 to 1.4920
Formula Weight 646.85
Solubility Solubility in water: soluble. Other solubilities: soluble in toluene, xylene, trichloroethylene,, ethylene glycol, ethyl ether, ethyl alcohol,, isopropyl alcohol, ethylene dichloride and most, other solvnets., insoluble in kerosene, miniral spirits.
Viscosity 240 mPa.s (25°C)
Boiling Point 270.0°C
Melting Point 6.0°C
Flash Point 274°C
Quantity 10L
Specific Gravity 1.067
Chemical Name or Material Triton™ X-100
Show More Show Less

GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
Avoid release to the environment.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger