Learn More
Triton™ X-100, ACROS Organics™
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Density | 1.0670g/mL |
UN Number | 3082 |
Merck Index | 15, 6850 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in toluene, xylene, trichloroethylene,, ethylene glycol, ethyl ether, ethyl alcohol,, isopropyl alcohol, ethylene dichloride and most, other solvnets., insoluble in kerosene, miniral spirits. |
Packaging | Glass Bottle |
Refractive Index | 1.4900 to 1.4920 |
Formula Weight | 646.85 |
Viscosity | 240 mPa.s (25°C) |
Boiling Point | 270.0°C |
Melting Point | 6.0°C |
Show More |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Harmful if swallowed.
GHS P Statement
Avoid release to the environment.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Danger