Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Triton™ X-100 Lysis Buffer with Glycerol, 2X, pH 7.4

$44.97 - $69.80

Specifications

Concentration 2X
Format Liquid
Description Colorless
pH 7.4
Product Type Cell Lysis Reagent
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ63866AK
View Documents Promotion Details
Thermo Scientific™
J63866AK
250mL
Each for $69.80
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ63866EQE
View Documents Promotion Details
Thermo Scientific™
J63866EQE
125mL
Each for $44.97
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2X
Colorless
Cell Lysis Reagent
Liquid
7.4
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit