Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triton X-100™, Reagent, Reagents

Catalog No. p-200021118
Click to view available options
Quantity:
250 mL
500 mL
1 L

Triton X-100™, Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula (CH3)3CCH2C(CH3)2C6H4(OCH2CH2)x
Molecular Weight (g/mol) 250.1933
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Octoxynol, Octoxinol
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 L
Packaging Poly Bottle
Specific Gravity 1.065
Chemical Name or Material Triton X-100™, Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.