Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triton™ X-405, ACROS Organics™
Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
Specifications
Density | 1.0800g/mL |
Merck Index | 15, 6850 |
Packaging | Glass bottle |
Refractive Index | 1.4360 to 1.4380 |
Formula Weight | 1968.47 |
Physical Form | Liquid |
Percent Purity | ≥65% |
Solubility | Solubility in water: soluble. |
Boiling Point | 120.0°C |
Color | Yellow |
Show More |
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
GHS P Statement:
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.