Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triton™ X-405, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
9002-93-1,7732-18-5
C16H26O2
250.382
MFCD01779734
JYCQQPHGFMYQCF-UHFFFAOYSA-N
5590
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
9002-93-1,7732-18-5
C16H26O2
250.382
MFCD01779734
JYCQQPHGFMYQCF-UHFFFAOYSA-N
5590
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
Density 1.0800g/mL
Merck Index 15, 6850
Packaging Glass bottle
Refractive Index 1.4360 to 1.4380
Formula Weight 1968.47
Physical Form Liquid
Percent Purity ≥65%
Solubility Solubility in water: soluble.
Boiling Point 120.0°C
Color Yellow
Melting Point -4.0°C
Flash Point >109°C
Quantity 250mL
Specific Gravity 1.08
Chemical Name or Material Triton™ X-405, 70% Solution in Water
Show More Show Less

GHS H Statement:
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.