Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trityl Tetrakis(pentafluorophenyl)borate, 97%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
Specifications
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Color | Yellow |
Quantity | 1g |
Assay Percent Range | 97% |
Formula Weight | 922.37 |
Physical Form | Powder |
Percent Purity | 97% |
Chemical Name or Material | Trityl tetrakis(pentafluorophenyl)borate |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning