Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trityl Tetrakis(pentafluorophenyl)borate, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
136040-19-2
C43H15BF20
922.37
MFCD03426981
TZOSNOQHGGONMD-UHFFFAOYSA-N
triphenylmethylium tetrakis perfluorophenyl borate,trityl tetrakis pentafluorophenyl borate,trityl tetrapentafluorophenyl borate,methylium, triphenyl-, tetrakis pentafluorophenyl borate 1-,triphenylcarbenium tetrakis pentafluorophenyl borate,methylium, triphenyl-, tetrakis 2,3,4,5,6-pentafluorophenyl borate 1-1:1,tetrakis 2,3,4,5,6-pentafluorophenyl boranuide; triphenylmethylium,methylium, triphenyl-, tetrakis pentafluorophenyl borate 1-1:1,tritylium tetrakis pentafluorophenyl borate,triphenylmethyliumtetrakis pentafluorophenyl borate
9832824
diphenylmethylbenzene;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3
136040-19-2
C43H15BF20
922.37
MFCD03426981
TZOSNOQHGGONMD-UHFFFAOYSA-N
triphenylmethylium tetrakis perfluorophenyl borate,trityl tetrakis pentafluorophenyl borate,trityl tetrapentafluorophenyl borate,methylium, triphenyl-, tetrakis pentafluorophenyl borate 1-,triphenylcarbenium tetrakis pentafluorophenyl borate,methylium, triphenyl-, tetrakis 2,3,4,5,6-pentafluorophenyl borate 1-1:1,tetrakis 2,3,4,5,6-pentafluorophenyl boranuide; triphenylmethylium,methylium, triphenyl-, tetrakis pentafluorophenyl borate 1-1:1,tritylium tetrakis pentafluorophenyl borate,triphenylmethyliumtetrakis pentafluorophenyl borate
9832824
diphenylmethylbenzene;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Yellow
Assay Percent Range 97%
Formula Weight 922.37
Physical Form Powder
Percent Purity 97%
Chemical Name or Material Trityl tetrakis(pentafluorophenyl)borate

GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning